missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZBTB1 Polyclonal specifically detects ZBTB1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ZBTB1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | zinc finger and BTB domain containing 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ZBTB1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?