missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZBP1 Recombinant Rabbit Monoclonal Antibody (4S9N3)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558195

 View more versions of this product

Product Code. 30284696

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAQAPADPGR EGHLEQRILQ VLTEAGSPVK LAQLVKECQA PKRELNQVLY RMKKELKVSL TSPATWCLGG TDPEGEGPAE LALSSPAERP QQHAATIPET PGPQFSQQRE EDIYRFLKDN GPQRALVIAQ ALGMRTAKDV NRDLYRMKSR HLLDMDEQSK AWTIYRPEDS.

Z-DNA binding protein 1 (ZBP1) belongs to a family of proteins that contain the Zalpha domain which binds specifically to left-handed Z-DNA and Z-RNA. ZBP1 was initially identified as a novel gene that was up-regulated in activated macrophages in mice bearing ascites tumors, suggesting that is may play a role in processes such as host response in neoplasia. More recent reports indicate that the cytosolic ZBP1 has can act like the toll-like receptor TLR9 by detecting cytosolic double-stranded (ds) DNA and trigger induction of type I interferon and other innate immune responses. It is thought that the binding of ZBP1 to dsDNA enhances its association with innate immune response proteins such as the IRF3 transcription factor and the serine/threonine kinase TBK1 (also known as NAK). Multiple isoforms of ZBP1 are known to exist.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZBP1
Recombinant Monoclonal
1.67 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q9H171
ZBP1
Recombinant protein (or fragment).
20 μL
Primary
Human
Antibody
IgG
ELISA, Immunoprecipitation
4S9N3
Unconjugated
ZBP1
2010010H03Rik; C20orf183; Dai; Dlm1; DLM-1; DNA-dependent activator of IFN-regulatory factors; DNA-dependent activator of IRFs; mZaDLM; RP4-718J7.1; Tumor stroma and activated macrophage protein DLM-1; Zbp1; Z-DNA binding protein 1; Z-DNA-binding protein 1
Rabbit
Protein A
RUO
81030
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.