missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBP1, Mouse anti-Human, Clone: 2C10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00081030-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: AQAPADPGREGHLEQRILQVLTEAGSPVKLAQLVKECQAPKRELNQVLYRMKKELKVSLTSPATWCLGGTDPEGEGPAELALSSPAKRPQQHAATIPETSpecifications
| ZBP1 | |
| Monoclonal | |
| Unconjugated | |
| Z-DNA binding protein 1 | |
| C20orf183/DAI/DLM-1/DLM1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2C10 | |
| In 1x PBS, pH 7.4 | |
| NM_030776 | |
| ZBP1 | |
| ZBP1 (NP_110403.1, 2 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 81030 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction