missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBP1/DLM-1/DAI Antibody (Zippy-1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80056AF350
This item is not returnable.
View return policy
Description
ZBP1/DLM-1/DAI Monoclonal antibody specifically detects ZBP1/DLM-1/DAI in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| ZBP1/DLM-1/DAI | |
| Monoclonal | |
| Alexa Fluor 350 | |
| C20orf183, chromosome 20 open reading frame 183, DAI, DLM1, DLM-1, DLM1dJ718J7.3, DNA-dependent activator of interferon regulatory factors, DNA-dependent activator of IRFs, Tumor stroma and activated macrophage protein DLM-1, ZBP1 Z-DNA binding protein 1, Z-DNA binding protein 1, Z-DNA-binding protein 1 | |
| Recombinant mouse ZBP1/DLM-1/DAI (aa 1-411) | |
| 0.1 mL | |
| Innate Immunity | |
| 81030 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunofluorescence | |
| Zippy-1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction