missing translation for 'onlineSavingsMsg'
Learn More

ZBED5 Antibody, Novus Biologicals™

Product Code. 18415969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415969 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415969 Supplier Novus Biologicals Supplier No. H00058486B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ZBED5 Polyclonal antibody specifically detects ZBED5 in Human samples. It is validated for Western Blot, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBED5
Applications Western Blot, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Chromatin Immunoprecipitation (ChIP)
Formulation PBS (pH 7.4)
Gene Alias zinc finger BED domain-containing protein 5, zinc finger, BED-type containing 5
Host Species Mouse
Immunogen ZBED5 (AAH38508.1, 1 a.a. - 693 a.a.) full-length human protein. MIAPILCILSYNFNTFAILNVYSKLTMFCTTNSLPMDLLLKQGSLKQEVESFCYQIVSESNDQKVGILQSEDKQLQPSVSKKSEGELSRVKFISNSNKITFSKKPKRRKYDESYLSFGFTYFGNRDAPHAQCVLCKKILSNSSLAPSKLRRHLETKHAAYKDKDISFFKQHLDSPENNKPPTPKIVNTDNESATEASYNVSYHIALSGEAHTIGELLIKPCAKDVVMRMFDEQYSKKIDAVQLSNSTVARRIKDLAADIEEELVCRLKICDGFSLQLDESADVSGLAVLLVFVRYRFNKSIEEDLLLCESLQSNATGEEIFNCINSFMQKHEIEWEKCVDVCSDASRAVDGKIAEAVTLIKYVAPESTSSHCLLYRHALAVKIMPTSLKNVLDQAVQIINYIKARPHQSRLLKILCEEMGAQHTALLLNTEVRWLSRGKVLVRLFELRRELLVFMDSAFRLSDCLTNSSWLLRLAYLADIFTKLNEVNLSMQGKNVTVFTVFDKMSSLLRKLEFWASSVEEENFDCFPTLSDFLTEINSTVDKDICSAIVQHLRGLRATLLKYFPVTNDNNAWVRNPFTVTVKPASLVARDYESLIDLTSDSQVKQNFSELSLNDFWSSLIQEYPSIARRAVRVLLPFATMHLCETGFSYYAATKTKYRKRLDAAPHMRIRLSNITPNIKRICDKKTQKHCSH
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58486
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.