missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBED2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | ZBED2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
ZBED2 Polyclonal specifically detects ZBED2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry.Specifications
| ZBED2 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_078784 | |
| 79413 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human ZBED2. The immunogen is located within the first 50 amino acids of ZBED2. Amino Acid Squence: DLEMKEEEEISETGE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| MGC10796, zinc finger BED domain-containing protein 2, zinc finger, BED domain containing 2, zinc finger, BED-type containing 2 | |
| ZBED2 | |
| IgG | |
| Affinity Purified | |
| This antibody is predicted to have no cross-reactivity to ZBED1 or ZBED3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title