missing translation for 'onlineSavingsMsg'
Learn More

ZBED2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18429402 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429402 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429402 Supplier Novus Biologicals Supplier No. NBP177349AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZBED2 Polyclonal antibody specifically detects ZBED2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBED2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias MGC10796, zinc finger BED domain-containing protein 2, zinc finger, BED domain containing 2, zinc finger, BED-type containing 2
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human ZBED2. The immunogen is located within the first 50 amino acids of ZBED2. Amino Acid Squence: DLEMKEEEEISETGE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79413
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.