missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zardaverine, Tocris Bioscience™
PDE3/4 inhibitor
£138.33 - £516.19
Chemical Identifiers
CAS | 101975-10-4 |
---|---|
Molecular Formula | C12H10F2N2O3 |
Molecular Weight (g/mol) | 268.22 |
InChI Key | HJMQDJPMQIHLPB-UHFFFAOYSA-N |
Synonym | zardaverine, zardaverina, zardaverinum, zardaverine inn, zardaverinum inn-latin, unii-tq358gwh6y, zardaverina inn-spanish, 6-4-difluoromethoxy-3-methoxy-phenyl-2h-pyridazin-3-one, 6-4-difluoromethoxy-3-methoxyphenyl-3 2h-pyridazinone, tq358gwh6y |
PubChem CID | 5723 |
ChEBI | CHEBI:46548 |
IUPAC Name | 3-[4-(difluoromethoxy)-3-methoxyphenyl]-1H-pyridazin-6-one |
SMILES | COC1=C(C=CC(=C1)C2=NNC(=O)C=C2)OC(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18015462
|
Tocris Bioscience
1046/10 |
10 mg |
£138.33
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30002152
|
Tocris Bioscience
1046/50 |
50 mg |
£516.19
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphodiesterase inhibitor, selective for PDE3 and 4 (IC50 values are 0.5 and 0.8 μM respectively).Chemical Identifiers
101975-10-4 | |
268.22 | |
zardaverine, zardaverina, zardaverinum, zardaverine inn, zardaverinum inn-latin, unii-tq358gwh6y, zardaverina inn-spanish, 6-4-difluoromethoxy-3-methoxy-phenyl-2h-pyridazin-3-one, 6-4-difluoromethoxy-3-methoxyphenyl-3 2h-pyridazinone, tq358gwh6y | |
CHEBI:46548 | |
COC1=C(C=CC(=C1)C2=NNC(=O)C=C2)OC(F)F |
C12H10F2N2O3 | |
HJMQDJPMQIHLPB-UHFFFAOYSA-N | |
5723 | |
3-[4-(difluoromethoxy)-3-methoxyphenyl]-1H-pyridazin-6-one |
Specifications
101975-10-4 | |
zardaverine, zardaverina, zardaverinum, zardaverine inn, zardaverinum inn-latin, unii-tq358gwh6y, zardaverina inn-spanish, 6-4-difluoromethoxy-3-methoxy-phenyl-2h-pyridazin-3-one, 6-4-difluoromethoxy-3-methoxyphenyl-3 2h-pyridazinone, tq358gwh6y | |
COC1=C(C=CC(=C1)C2=NNC(=O)C=C2)OC(F)F | |
268.22 | |
CHEBI:46548 | |
>98% |
C12H10F2N2O3 | |
HJMQDJPMQIHLPB-UHFFFAOYSA-N | |
3-[4-(difluoromethoxy)-3-methoxyphenyl]-1H-pyridazin-6-one | |
5723 | |
268.22 | |
Zardaverine |
Safety and Handling
Recommended Storage : Store at Room Temperature