missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZAR1L Polyclonal specifically detects ZAR1L in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ZAR1L |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | ZAR1-like protein, ZAR2, zygote arrest 1-like |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human ZAR1L |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?