missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZAR1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559514
This item is not returnable.
View return policy
Description
Immunogen sequence: PRTLSSCSPW DGRDPQEPLP ACGVTSPATG RRGLIRLRRD GDEAESKALP GPAEASQPQP PSRRSGADRQ EEPGQLEESG EKDAPCPQET KSKQVPGDAA SEPLRRPNFQ F Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 30%.
This gene encodes a member of the ZAR1 family that is predominantly expressed in oocytes and early embryos. The protein may function as an RNA regulator in early embryos.
Specifications
| ZAR1L | |
| Polyclonal | |
| Unconjugated | |
| ZAR1L | |
| XM_359419; Z3CXXC7; ZAR1L; ZAR1-like protein; ZAR2; zinc finger, 3CxxC-type 7; zygote arrest 1-like; zygotic arrest 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 646799 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NP61 | |
| ZAR1L | |
| Recombinant protein corresponding to Human ZAR1L. Recombinant protein control fragment (Product #RP-98011). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction