missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZAP-70, His Tag Recombinant Protein
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Protein Form: Full Length.
ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins. ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling.
Specifications
Specifications
| Accession Number | P43403 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 25mM HEPES with 150mM NaCl, 0.5mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 7535 |
| Molecular Weight (g/mol) | 70.1 kDa |
| Name | Human ZAP-70, His Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction