Learn More
Invitrogen™ Zap-70 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596093
Description
Immunogen sequence: MPDPAAHLPF FYGSISRAEA EEHLKLAGMA DGLFLLRQCL RSLGGYVLSL VHDVRFHHFP IERQLNGTYA IAGGKAHCGP AELCEFYSRD PDGLPCNLRK PCNRPSGLEP QPGVFDCLRD AMVRDYVRQT WKLEGEALEQ AIISQAPQVE KLIATTAHER MPWYHSSLTR EEAERKLYSG AQTDGKFLLR PRKEQGTYAL SLIYGKTVYH YLISQDKAGK YCIPEGTKFD TLWQLVEYLK LKADGLIYCL KEACPNSSAS NASGAAAPTL PAHPSTLTHP QRRIDTLNSD GYTPEPARIT; Positive Samples: SW480, U251, Mouse brain, Mouse spleen; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein.
ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins. ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling.
Specifications
| Zap-70 | |
| Polyclonal | |
| Unconjugated | |
| ZAP70 | |
| 70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; ADMIO2; EC 2.7.10.2; FLJ17670; FLJ17679; I79_004087; IMD48; kinase ZAP70; mrtle; mur; Selective T cell defect; Srk; STCD; STD; syk-related protein tyrosine kinase; syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; Tyrosine-protein kinase ZAP-70;TZK; TZK; ZA70; Zap70; ZAP-70; Zeta chain associated protein kinase 70kDa; zeta chain of T cell receptor associated protein kinase; zeta chain of T cell receptor associated protein kinase 70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase; zeta-chain (TCR) associated protein kinase 70; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22637, 301348, 7535 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43403, P43404 | |
| ZAP70 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZAP70 (NP_0010702). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.