missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ZAP 180013

Product Code. 18797916
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18797916 50 mg 50mg
18705756 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18797916 Supplier R&D Systems™ Supplier No. 6821/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Zap70 inhibitor; inhibits interaction with ITAMs

ZAP 180013 is a zeta-chain-associated protein kinase 70 (Zap70) inhibitor (IC50 = 1.8 μM in fluorescence polarization assay). Inhibits the interaction of Zap70 SH2 domain with immunoreceptor tyrosine-based activation motif (ITAMs), within the T-cell antigen receptor (TCR) signaling pathway.
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 5-Chloro-N-[(4-chlorophenyl)methyl]-2-(ethylsulfonyl)-N-(2-furanylmethyl)-4-pyrimidinecarboxamide
CAS 873080-25-2
Quantity 50 mg
Product Line Tocris
Molecular Formula C19H17Cl2N3O4S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.