missing translation for 'onlineSavingsMsg'
Learn More

Labcon ZAP™ 1000 μL Extended Length Aerosol Filter Pipette Tips
GREENER_CHOICE

Product Code. 16643942
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Rack
Refill
Color:
Clear
Natural
Unit Size:
Pack of 1000
Pack of 200
Pack of 576
Pack of 768
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format Color unitSize
16643942 Sterile Rack Clear Pack of 576
16603942 Sterile Bagged Clear Pack of 200
16613942 Sterile Refill Clear Pack of 768
16623942 Non-sterile Bagged Clear Pack of 1000
16633942 Sterile Rack Natural Pack of 576
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16643942 Supplier Labcon Supplier No. 10579650169
Add to basket
Add to basket

Fits Most Popular 1000 μL Pipettors

  • Pipette Tip
  • Fits Most Popular 1000 μL Pipettors
  • Patented robotically inserted endotoxin free filter
  • Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Made in USA

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Length (English) 3.50 in.
Length (Metric) 88.9 mm
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Clean Claims Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
For Use With (Application) Liquid Handing
Sterilization Method E-Beam
Format Rack
Tip Style UltraFine
Packaging 96 Tips/Rack, 6 Racks/Pk., 8 Pk./Cs.
Quantity 96 tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Product Line ZAP
Material Polypropylene Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.