missing translation for 'onlineSavingsMsg'
Learn More

Zamicastat, MedChemExpress

Product Code. 16466767
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466767

Brand: Medchem Express HY106004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zamicastat (BIA 5-1058) is a dopamine β-hydroxylase (DBH) inhibitor and can cross the blood-brain barrier (BBB) to cause central as well as peripheral effects. Zamicastat is also a concentration-dependent dual P-gp and BCRP inhibitor with IC50 values of 73.8 μM and 17.0 μM, respectively. Zamicastat reduces high blood pressure.

Zamicastat (BIA 5-1058) is a dopamine β-hydroxylase (DBH) inhibitor and can cross the blood-brain barrier (BBB) to cause central as well as peripheral effects. Zamicastat is also a concentration-dependent dual P-gp and BCRP inhibitor with IC50 values of 73.8 μM and 17.0 μM, respectively. Zamicastat reduces high blood pressure.

Specifications

Chemical Name or Material Zamicastat
CAS 1080028-80-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H21F2N3OS
Quantity 5 mg
Synonym BIA 5-1058
Solubility Information DMSO : 150 mg/mL (373.63 mM; Need ultrasonic)
SMILES S=C1N([C@@H]2CC3=CC(F)=CC(F)=C3OC2)C(CCNCC4=CC=CC=C4)=CN1
Molecular Weight (g/mol) 401.47
Formula Weight 401.47
Percent Purity 95.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.