missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Zalutumumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559402
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Zalutumumab is a fully human IgG1 monoclonal antibody designed to bind with selectivity to the epidermal growth factor receptor (EGFR). Zalutumumab has been investigated for the treatment of Squamous Cell Cancer and Head and Neck Cancer.
Specifications
| Zalutumumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HuMax-EGFr | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction