missing translation for 'onlineSavingsMsg'
Learn More

Zaltidine, MedChemExpress

Product Code. 16499177
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499177

Brand: Medchem Express HY15541

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zaltidine(CP-57361) is a H2-receptor antagonist, which has the antisecretory action.

Zaltidine(CP-57361) is a H2-receptor antagonist, which has the antisecretory action.

Specifications

Chemical Name or Material Zaltidine
Physical Form Solid
CAS 85604-00-8
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C8H10N6S
Synonym CP-57361
Solubility Information H2O : 7.69 mg/mL (34.60 mM; Need ultrasonic)
SMILES NC(NC1=NC(C2=CN=C(C)N2)=CS1)=N
Molecular Weight (g/mol) 222.27
Formula Weight 222.27
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.