missing translation for 'onlineSavingsMsg'
Learn More

Zaldaride maleate, MedChemExpress

Product Code. 16486727
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486727 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16486727 Supplier Medchem Express Supplier No. HY105118A5mg
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Zaldaride maleate (CGS-9343B) is a potent, orally active and selective inhibitor of calmodulin. Zaldaride maleate (CGS-9343B) inhibits CaM (calmodulin)-stimulated cAMP phosphodiesterase activity, with an IC50 of 3.3 nM. Zaldaride maleate (CGS-9343B) prevents estrogen-induced transcription activation by ER, reversibly blocks voltage-activated Na+, Ca2+ and K+ currents in PC12 cells and inhibits nAChR.

Zaldaride maleate (CGS-9343B) is a potent, orally active and selective inhibitor of calmodulin. Zaldaride maleate (CGS-9343B) inhibits CaM (calmodulin)-stimulated cAMP phosphodiesterase activity, with an IC50 of 3.3 nM. Zaldaride maleate (CGS-9343B) prevents estrogen-induced transcription activation by ER, reversibly blocks voltage-activated Na+, Ca2+ and K+ currents in PC12 cells and inhibits nAChR.

Specifications

Chemical Name or Material Zaldaride maleate
CAS 109826-27-9
Color White
Purity Grade Notes Research
Molecular Formula C30H32N4O6
Quantity 5 mg
Synonym CGS-9343B KW 5617
SMILES O=C(O)/C=C\C(O)=O.CC1(C2=CC=CN2C3=CC=CC=C3CO1)CN4CCC(N5C6=C(C=CC=C6)NC5=O)CC4
Molecular Weight (g/mol) 544.6
Formula Weight 544.6
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.