missing translation for 'onlineSavingsMsg'
Learn More

Zalcitabine, TRC

Product Code. 30449288
Change view
Click to view available options
Quantity:
10 mg
25 mg
1 g
Unit Size:
25mg
Product Code. Quantity unitSize
30449288 25 mg 25mg
30453068 1 g -
30451009 10 mg -
3 options
This item is not returnable. View return policy

Product Code. 30449288

Brand: Toronto Research Chemicals TRCZ14000025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7481-89-2
Formula Weight 211.0957
InChI Formula InChI=1S/C9H13N3O3/c10-7-3-4-12(9(14)11-7)8-2-1-6(5-13)15-8/h3-4,6,8,13H,1-2,5H2,(H2,10,11,14)/t6-,8+/m0/s1
IUPAC Name 4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Molecular Formula C9 H13 N3 O3
Molecular Weight (g/mol) 211.22
SMILES NC1=NC(=O)N(C=C1)[C@H]2CC[C@@H](CO)O2
Synonym Cytidine, 2',3'-dideoxy-, 2',3'-Dideoxycytidine, D 2C, Dideoxycytidine, Hivid, NSC 606170, Ro 24-2027/000, Zalcitabine, ddC
Chemical Name or Material Zalcitabine

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.