missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14945-1-AP-20UL
This item is not returnable.
View return policy
Description
MLKLAK is a member of the MAPKKK family of signal transduction molecules. It possesses an N-terminal kinase catalytic domain, followed by a leucine zipper motif and a sterile-alpha motif (SAM). This magnesium-binding protein forms homodimers and is located in the cytoplasm. The protein mediates gamma radiation signaling leading to cell cycle arrest and activity of this protein plays a role in cell cycle checkpoint regulation in cells. The protein also has pro-apoptotic activity.Specifications
| ZAK | |
| Polyclonal | |
| Unconjugated | |
| MAP3K20 | |
| AZK, HCCS 4, MLK related kinase, MLK7, mlklak, MLT, MLTK, MRK, ZAK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311743, 51776, 65964 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ESL4, Q9NYL2 | |
| Map3k20 | |
| ZAK Fusion Protein Ag6747 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction