missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZAK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579630
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse thymus tissue, human PANC-1 whole cell.
ZAK is a leucine zipper-containing mitogen-activated protein related to the MLKs in both sequence and function. Like the MLKs and other MAP3Ks, ZAK activates MAP2Ks. ZAK is a member of the MAPKKK family of signal transduction molecules and encodes a protein with an N-terminal kinase catalytic domain, followed by a leucine zipper motif and a sterile-alpha motif. This magnesium-binding protein forms homodimers and is located in the cytoplasm. The protein mediates gamma radiation signaling leading to cell cycle arrest and activity of this protein plays a role in cell cycle checkpoint regulation in cells. The protein also has pro-apoptotic activity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| ZAK | |
| Polyclonal | |
| Unconjugated | |
| MAP3K20 | |
| AV006891; AZK; B230120H23Rik; HCCS4; HCCS-4; Human cervical cancer suppressor gene 4 protein; leucine zipper- and sterile alpha motif kinase ZAK; Leucine zipper- and sterile alpha motif-containing kinase; Map3k20; Mitogen-activated protein kinase kinase kinase 20; Mitogen-activated protein kinase kinase kinase MLT; mixed lineage kinase 7; mixed lineage kinase with a leucine zipper and a sterile alpha motif; mixed lineage kinase-related kinase; mixed lineage kinase-related kinase MRK-beta; MLK7; mlklak; MLK-like mitogen-activated protein triple kinase; MLK-related kinase; MLT; MLTK; MLTKalpha; MLTKbeta; MRK; pk; RGD1561394; SFMMP; sterile alpha motif and leucine zipper containing kinase AZK; sterile alpha motif- and leucine zipper-containing kinase AZK; sterile-alpha motif and leucine zipper containing kinase AZK; ZAK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 311743, 51776, 65964 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q9ESL4, Q9NYL2 | |
| MAP3K20 | |
| E. coli-derived human MAP3K20 recombinant protein (Position: F276-E511). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction