missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35439-20ul
This item is not returnable.
View return policy
Description
ZAK Polyclonal antibody specifically detects ZAK in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| ZAK | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| AZK, EC 2.7.11, EC 2.7.11.25, HCCS-4, Human cervical cancer suppressor gene 4 protein, Leucine zipper- and sterile alpha motif-containing kinase, mitogen-activated protein kinase kinase kinase MLT, mixed lineage kinase 7, mixed lineage kinase with a leucine zipper and a sterile alpha motif, Mixed lineage kinase-related kinase, mixed lineage kinase-related kinase MRK-beta, MLK7, mLklak, MLK-like mitogen-activated protein triple kinase, MLK-related kinase, MLT, MLTK, MRKcervical cancer suppressor gene 4 protein, pk, protein kinase, sterile alpha motif and leucine zipper containing kinase AZK, Sterile alpha motif- and leucine zipper-containing kinase AZK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-312 of human ZAK (NP_598407.1).,, Sequence:, EEMDMDHIMTWATDVAKGMHYLHMEAPVKVIHRDLKSRNVVIAADGVLKICDFGASRFHNHTTHMSLVGTFPWMAPEVIQSLPVSETCDTYSYGVVLWEMLTREVPFKGLEGLQVAWLVVEKNERLTIPSSCPRSFAELLHQCWEADAKKRPSFKQIISILESMSNDTSLPDKCNSFLHNKAEWRCEIEATLERLKKLERDLSFKEQELKERE | |
| 20 μL | |
| Cell Cycle and Replication | |
| 51776 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction