missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAK Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18882G
This item is not returnable.
View return policy
Description
ZAK Polyclonal antibody specifically detects ZAK in Human samples. It is validated for Western Blot
Specifications
| ZAK | |
| Polyclonal | |
| Western Blot | |
| AZK, EC 2.7.11, EC 2.7.11.25, HCCS-4, Human cervical cancer suppressor gene 4 protein, Leucine zipper- and sterile alpha motif-containing kinase, mitogen-activated protein kinase kinase kinase MLT, mixed lineage kinase 7, mixed lineage kinase with a leucine zipper and a sterile alpha motif, Mixed lineage kinase-related kinase, mixed lineage kinase-related kinase MRK-beta, MLK7, mlklak, MLK-like mitogen-activated protein triple kinase, MLK-related kinase, MLT, MLTK, MRKcervical cancer suppressor gene 4 protein, pk, protein kinase, sterile alpha motif and leucine zipper containing kinase AZK, Sterile alpha motif- and leucine zipper-containing kinase AZK | |
| The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human sterile alpha motif and leucine zipper containing kinase AZK using the numbering given in entry NP_057737.2 (GeneID 51776). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51776 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction