missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAG Antibody (Zagy-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80083
This item is not returnable.
View return policy
Description
ZAG Monoclonal specifically detects ZAG in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ZAG | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 563 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| Zagy-1 | |
| Unconjugated | |
| PBS containing 10% glycerol | |
| AZGP1 | |
| Recombinant human ZAG. | |
| 100 μg | |
| Primary | |
| Recognizes human ZAG. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction