missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Zafirlukast

Product Code. 16621862 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16621862

Brand: Thermo Scientific Chemicals 463290010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 107753-78-6 | C31H33N3O6S | 575.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 107753-78-6
Molecular Formula C31H33N3O6S
MDL Number MFCD00864775
Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: slightly soluble in acetone, soluble in dimethylformamide and pyridine, insoluble in methanol, toluene, ethyl acetate & acetonitrile
InChI Key YEEZWCHGZNKEEK-UHFFFAOYSA-N
SMILES COC1=CC(=CC=C1CC1=CN(C)C2=CC=C(NC(=O)OC3CCCC3)C=C12)C(=O)NS(=O)(=O)C1=CC=CC=C1C
IUPAC Name cyclopentyl N-[3-({2-methoxy-4-[(2-methylbenzenesulfonyl)carbamoyl]phenyl}methyl)-1-methyl-1H-indol-5-yl]carbamate
Molecular Weight (g/mol) 575.68
Formula Weight 575.68
Chemical Name or Material Zafirlukast
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.