missing translation for 'onlineSavingsMsg'
Learn More

ZA20D3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

£222.00 - £444.00

Specifications

Antigen ZA20D3
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18465831
View Documents
Novus Biologicals
NBP1-90060-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18719983
View Documents
Novus Biologicals
NBP1-90060
£444.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZA20D3 Polyclonal specifically detects ZA20D3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ZA20D3
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
54469
This antibody was developed against Recombinant Protein corresponding to amino acids:SSNGRISPPATSVSSLSESLPVQCTDGSVPEAQSALDSTSSSMQPSPVSNQSLLSESVASSQLDSTSVDK
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Human
Associated with PRK1 protein, AWP1Zinc finger A20 domain-containing protein 3, protein associated with PRK1, ZA20D3AN1-type zinc finger protein 6, ZFAND5B, zinc finger, A20 domain containing 3, zinc finger, AN1-type domain 6
ZFAND6
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.