missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Z-Val-Ala-Asp (OMe)-FMK Inhibitor

Product Code. p-5538765 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 18172444

Brand: Novus Biologicals™ NBP229392

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in research applications

TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Store at -20°C. Avoid freeze-thaw cycles
Quantity 1 mg
Product Type Z-Val-Ala-Asp (OMe)-FMK Inhibitor
Molecular Weight (g/mol) 467
Inhibitors Z-Val-Ala-Asp (OMe)-FMK
Form Solid

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.