missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Z-VAD(OMe)-Fluromethylketone Inhibitor

Product Code. p-8015227 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
3 mg
5 mg
Unit Size:
1mg
3mg
5mg
This item is not returnable. View return policy

Product Code. 11404906

Brand: MP Biomedicals™ 03FK00901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Z-VAD(OMe)-FMK inhibitor synthesized using proprietary technology is cell permeable and irreversibly binds to activated Caspases to block apoptosis.

Z-VAD(OMe)-FMK is an irreversible and cell permeable broad-spectrum Caspase Inhibitor. It has been shown to inhibit Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine).

TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage 4°C
Quantity 1 mg
Product Type Z-Val-Ala-Asp-(OMe)-Fluromethylketone Inhibitor
Molecular Weight (g/mol) 467.487
Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.