missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Thiothixene, TRC

Product Code. 30452551
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30452551 25 mg 25mg
30449528 100 mg -
30449771 5 mg -
3 options
This item is not returnable. View return policy

Product Code. 30452551

Brand: Toronto Research Chemicals TRCT37527525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3313-26-6
Formula Weight 443.1701
InChI Formula InChI=1S/C23H29N3O2S2/c1-24(2)30(27,28)18-10-11-23-21(17-18)19(20-7-4-5-9-22(20)29-23)8-6-12-26-15-13-25(3)14-16-26/h4-5,7-11,17H,6,12-16H2,1-3H3/b19-8-
IUPAC Name (9Z)-N,N-dimethyl-9-[3-(4-methylpiperazin-1-yl)propylidene]thioxanthene-2-sulfonamide
Molecular Formula C23 H29 N3 O2 S2
Molecular Weight (g/mol) 443.63
SMILES CN(C)S(=O)(=O)c1ccc2Sc3ccccc3\C(=C\CCN4CCN(C)CC4)\c2c1
Synonym 9H-Thioxanthene-2-sulfonamide, N,N-dimethyl-9-[3-(4-methyl-1-piperazinyl)propylidene]-, (9Z)-, 9H-Thioxanthene-2-sulfonamide, N,N-dimethyl-9-[3-(4-methyl-1-piperazinyl)propylidene]-, (Z)-, Thioxanthene-2-sulfonamide, N,N-dimethyl-9-[3-(4-methyl-1-piperazinyl)propylidene]-, (Z)- (8CI), Thiothixene, Tiotixene, cis-Thiothixene
Chemical Name or Material Thiothixene

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.