missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)-Thiothixene-d8, TRC
Click to view available options
Quantity:
0.5 mg
2.5 mg
10 mg
Unit Size:
0.5mg
10mg
2.5mg
Specifications
Specifications
| Chemical Name or Material | (Z)-Thiothixene-d8 |
| Molecular Formula | C23H21D8N3O2S2 |
| Quantity | 0.5 mg |
| Synonym | N,N-Dimethyl-9-[3-(4-methyl-1-piperazinyl)propylidene]- thioxanthene-2-sulfonamide-d8 (7CI,8CI), N,N-Dimethyl-9-[3-(4-methyl-1- |
| InChI Formula | InChI=1S/C23H29N3O2S2/c1-24(2)30(27,28)18-10-11-23-21(17-18)19(20-7-4-5-9-22(20)29-23)8-6-12-26-15-13-25(3)14-16-26/h4-5,7-11,17H,6,12-16H2,1-3H3/b19-8+/i13D2,14D2,15D2,16D2 |
| SMILES | O=[S](C1=CC2=C(C=C1)SC3=CC=CC=C3/C2=C\CCN4C([2H])([2H])C([2H])([2H])N(C)C([2H])([2H])C4([2H])[2H])(N(C)C)=O |
| IUPAC Name | (E)-N,N-dimethyl-9-(3-(4-methylpiperazin-1-yl-2,2,3,3,5,5,6,6-d8)propylidene)-9H-thioxanthene-2-sulfonamide |
| Molecular Weight (g/mol) | 451.67 |
| Formula Weight | 451.22 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction