missing translation for 'onlineSavingsMsg'
Learn More

[(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade), TRC

Product Code. 30455521
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455521 1 g 1g
30450621 5 g 5g
30451824 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455521 Supplier Toronto Research Chemicals Supplier No. TRCZ1927001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25383-06-6
Molecular Formula C3H7O3P
Molecular Weight (g/mol) 122.06
Synonym Phosphonic acid, P-(1Z)-1-propen-1-yl- (ACI), P-(1Z)-1-Propen-1-ylphosphonic acid (ACI), Phosphonic acid, (1Z)-1-propenyl- (9CI), Phosphonic acid, 1-propenyl-, (Z)- (ZCI), Phosphonic acid, propenyl-, (Z)- (8CI), (cis-1-Propenyl)phosphonic acid, [(Z)-1-Propenyl]phosphonic acid, cis-Propenylphosphonic acid
IUPAC Name [(Z)-prop-1-enyl]phosphonic acid
SMILES C\C=C/P(=O)(O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H7O3P/c1-2-3-7(4,5)6/h2-3H,1H3,(H2,4,5,6)/b3-2-
Formula Weight 122.0133
Chemical Name or Material [(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.