missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Z-LE(OMe)HD(OMe)-FMK (Caspase 9 Inhibitor)
Shop All Bio Techne Products
Click to view available options
Unit Size:
1mg
5mg
Quantity:
5 mg
Specifications
Specifications
| Color | Tan |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction