missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Heptyl Hexadec-9-enoate, TRC

Product Code. 30395826
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395826 50 mg 50mg
30404574 250 mg 250mg
30395238 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395826 Supplier Toronto Research Chemicals Supplier No. TRCH28319050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Z)-Heptyl Hexadec-9-enoate
Analyte or Component Names (Z)-Heptyl Hexadec-9-enoate
Shipping Condition Room Temperature
Molecular Formula C23 H44 O2
InChI Formula InChI=1S/C23H44O2/c1-3-5-7-9-10-11-12-13-14-15-16-17-19-21-23(24)25-22-20-18-8-6-4-2/h11-12H,3-10,13-22H2,1-2H3/b12-11-
Solution Type Neat
SMILES CCCCCCCOC(=O)CCCCCCC\C=C/CCCCCC
IUPAC Name heptyl (Z)-hexadec-9-enoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 352.594
Quantity 50 mg
Formula Weight 352.334
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.