missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Gemifloxacin Mesilate, TRC

Product Code. 30240830
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240830 100 mg 100mg
30240682 10 mg 10mg
30240653 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240830 Supplier Toronto Research Chemicals Supplier No. TRCG336000100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210353-53-0
Molecular Formula C18 H20 F N5 O4 . C H4 O3 S
Molecular Weight (g/mol) 485.49
Synonym 1,8-Naphthyridine-3-carboxylic acid, 7-[(4Z)-3-(aminomethyl)-4-(methoxyimino)-1-pyrrolidinyl]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-, monomethanesulfonate, Factive, Floxguard, Gemifloxacin mesylate, Gemixa, LB 20304a, SB 265805S, Gemifloxacin Mesilate
IUPAC Name 7-[(4Z)-3-(aminomethyl)-4-methoxyiminopyrrolidin-1-yl]-1-cyclopropyl-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid;methanesulfonic acid
SMILES CO\N=C\1/CN(CC1CN)c2nc3N(C=C(C(=O)O)C(=O)c3cc2F)C4CC4.CS(=O)(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H20FN5O4.CH4O3S/c1-28-22-14-8-23(6-9(14)5-20)17-13(19)4-11-15(25)12(18(26)27)7-24(10-2-3-10)16(11)21-17;1-5(2,3)4/h4,7,9-10H,2-3,5-6,8,20H2,1H3,(H,26,27);1H3,(H,2,3,4)/b22-14+;
Formula Weight 485.138
Chemical Name or Material Gemifloxacin Mesilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.