missing translation for 'onlineSavingsMsg'
Learn More

Z-FA-FMK, TRC

Product Code. 30449538
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449538 5 mg -
30457201 1 mg 1mg
30453832 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449538 Supplier Toronto Research Chemicals Supplier No. TRCZ3500005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197855-65-5
Molecular Formula C21H23FN2O4
Molecular Weight (g/mol) 386.42
Synonym N-[(1S)-2-[(3-Fluoro-1-methyl-2-oxopropyl)amino]-2-oxo-1-(phenylmethyl)ethyl]-carbamic acid Phenylmethyl Ester, [1(S)]-[2-[(3-Fluoro-1-methyl-2-oxopropyl)amino]-2-oxo-1-(phenylmethyl)ethyl]-carbamic acid Phenylmethyl Ester
IUPAC Name benzyl N-[(2S)-1-[(4-fluoro-3-oxobutan-2-yl)amino]-1-oxo-3-phenylpropan-2-yl]carbamate
SMILES CC(NC(=O)[C@H](Cc1ccccc1)NC(=O)OCc2ccccc2)C(=O)CF

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H23FN2O4/c1-15(19(25)13-22)23-20(26)18(12-16-8-4-2-5-9-16)24-21(27)28-14-17-10-6-3-7-11-17/h2-11,15,18H,12-14H2,1H3,(H,23,26)(H,24,27)/t15?,18-/m0/s1
Formula Weight 386.1642
Chemical Name or Material Z-FA-FMK
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.