missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Doxepin Hydrochloride, TRC

Product Code. 30372114
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372114 100 mg
30383797 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30372114 Supplier Toronto Research Chemicals Supplier No. TRCD550015100MG
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25127-31-5
Molecular Formula C19H22ClNO
Molecular Weight (g/mol) 315.84
Synonym (3Z)-3-(Dibenz[b,e]oxepin-11(6H)-ylidene)-N,N-dimethyl-1-propanamine Hydrochloride, (Z)-3-(Dibenz[b,e]oxepin-11(6H)-ylidene)-N,N-dimethyl-1-propanamine Hydrochloride, (Z)-N,N-dimethyldibenz[b,e]oxepin-delta11(6H),gamma-propylamine Hydrochloride, Dibenz[b,e]oxepin 1-Propanamine deriv.
SMILES Cl.CN(C)CC\C=C/1\c2ccccc2COc3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H21NO.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11-;
Formula Weight 315.139
Chemical Name or Material (Z)-Doxepin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.