missing translation for 'onlineSavingsMsg'
Learn More

Z-Diniconazole, TRC

Product Code. 30369107
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369107 10 mg 10mg
30381610 50 mg 50mg
30377476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369107 Supplier Toronto Research Chemicals Supplier No. TRCD47982110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Z)-Diniconazole
CAS 76714-16-4
Analyte or Component Names (Z)-Diniconazole
Shipping Condition Room Temperature
Molecular Formula C15 H17 Cl2 N3 O
Synonym 1H-1,2,4-Triazole-1-ethanol, β-[(2,4-dichlorophenyl)methylene]-α-(1,1-dimethylethyl)-, (βZ)-, 1H-1,2,4-Triazole-1-ethanol, β-[(2,4-dichlorophenyl)methylene]-α-(1,1-dimethylethyl)-, (Z)-, (βZ)-β-[(2,4-Dichlorophenyl)methylene]-α-(1,1-dimethylethyl)-1H-1,2,4-triazole-1-ethanol, (Z)-1-(2,4-Dichlorophenyl)-2-(1,2,4-triazol-1-yl)-4,4-dimethyl-1-penten-3-ol, Z-SO 8, (Z)-Diniconazole
InChI Formula InChI=1S/C15H17Cl2N3O/c1-15(2,3)14(21)13(20-9-18-8-19-20)6-10-4-5-11(16)7-12(10)17/h4-9,14,21H,1-3H3/b13-6-
Solution Type Neat
SMILES CC(C)(C)C(O)\C(=C\c1ccc(Cl)cc1Cl)\n2cncn2
IUPAC Name (Z)-1-(2,4-dichlorophenyl)-4,4-dimethyl-2-(1,2,4-triazol-1-yl)pent-1-en-3-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 326.221
Quantity 10 mg
Formula Weight 325.075
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.