missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (Z)-4-Hydroxytamoxifen

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
(Z)-4-Hydroxytamoxifen is an estrogen receptor antagonist (IC50 = 27 μM). (Z)-4-Hydroxytamoxifen is a Tamoxifen (Cat. No. 0999) metabolite; that exhibits greater potency than the parent compound. (Z)-4-Hydroxytamoxifen Also activates intein-linked inactive Cas9, reducing off-target CRISPR-mediated gene editing; system has ∽25-fold higher specificity than wtCas9. (Z)-4-Hydroxytamoxifen binds to voltage-gated sodium channels, near the intracellular gate and inhibits the sodium current by delaying channel recovery from the inactivated state (IC50 values are 297 nM and 2.1 μM for NavM and human Nav, respectively in HEK cells); this inhibition is independent of estrogen receptor modulation. Chemotherapeutic agent.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 387.51 |
| Purity | >98% |
| Molecular Weight (g/mol) | 387.523 |
| Chemical Name or Material | (Z)-4-Hydroxytamoxifen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction