missing translation for 'onlineSavingsMsg'
Learn More

(Z)-4-Hydroxytamoxifen, Thermo Scientific™

Product Code. 17740659 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17740659 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17740659 Supplier Thermo Scientific Acros Supplier No. 469501000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material (Z)-4-Hydroxytamoxifen
CAS 68047-06-3
Quantity 100 mg
Melting Point 137°C to 143°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C26 H29 N O2
Physical Form Crystalline Powder
Color White to Almost-white
Purity ≥98.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.