missing translation for 'onlineSavingsMsg'
Learn More

(Z)-4-((5-Hydroxy-3-methyl-1-phenyl-1H-pyrazol-4-yl)imino)-3-methyl-1-phenyl-1H-pyrazol-5(4H)-one, TRC

Product Code. 30389625
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389625 10 mg -
30398420 50 mg 50mg
30389034 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389625 Supplier Toronto Research Chemicals Supplier No. TRCH12173010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62094-95-5
Molecular Formula C20H17N5O2
Molecular Weight (g/mol) 359.38
Synonym (4Z)-2,4-Dihydro-4-[(5-hydroxy-3-methyl-1-phenyl-1H-pyrazol-4-yl)imino]-5-methyl-2-phenyl-3H-pyrazol-3-one
IUPAC Name (4Z)-5-methyl-4-[(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)imino]-2-phenylpyrazol-3-one
SMILES CC1=NN(C(=O)C1\N=C/2\C(=NN(C2=O)c3ccccc3)C)c4ccccc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H17N5O2/c1-13-17(19(26)24(22-13)15-9-5-3-6-10-15)21-18-14(2)23-25(20(18)27)16-11-7-4-8-12-16/h3-12,17H,1-2H3/b21-18-
Formula Weight 359.1382
Chemical Name or Material (Z)-4-((5-Hydroxy-3-methyl-1-phenyl-1H-pyrazol-4-yl)imino)-3-methyl-1-phenyl-1H-pyrazol-5(4H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.