missing translation for 'onlineSavingsMsg'
Learn More

(Z)-4-(2,4-Difluorobenzoyl)piperidine Oxime, TRC

Product Code. 30372023
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372023 25 mg
30388779 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30372023 Supplier Toronto Research Chemicals Supplier No. TRCD44607025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 691007-05-3
Molecular Formula C12 H14 F2 N2 O
Molecular Weight (g/mol) 240.2492
Synonym Methanone, (2,4-difluorophenyl)-4-piperidinyl-, oxime, (1Z)-, (Z)-(2,4-Difluorophenyl)-4-piperidinylmethanone oxime
IUPAC Name (NZ)-N-[(2,4-difluorophenyl)-piperidin-4-ylmethylidene]hydroxylamine
SMILES O\N=C(\C1CCNCC1)/c2ccc(F)cc2F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H14F2N2O/c13-9-1-2-10(11(14)7-9)12(16-17)8-3-5-15-6-4-8/h1-2,7-8,15,17H,3-6H2/b16-12-
Formula Weight 240.1074
Chemical Name or Material (Z)-(2,4-Difluorophenyl)-(4-piperidyl)methanone Oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.