missing translation for 'onlineSavingsMsg'
Learn More

(Z)-4-[(1,3-Dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]4-amino]oxy]methyl]-benzoic acid, TRC

Product Code. 30385103
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385103 10 mg
30377240 1 mg
30388367 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385103 Supplier Toronto Research Chemicals Supplier No. TRCD54508010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149054-57-9
Molecular Formula C20 H19 N3 O4
Molecular Weight (g/mol) 365.3826
Synonym Benzoic acid, 4-[[[[(1,3-dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]amino]oxy]methyl]-, (Z)- (9CI)
IUPAC Name 4-[[(Z)-(1,3-dimethyl-5-phenoxypyrazol-4-yl)methylideneamino]oxymethyl]benzoic acid
SMILES Cc1nn(C)c(Oc2ccccc2)c1\C=N/OCc3ccc(cc3)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H19N3O4/c1-14-18(19(23(2)22-14)27-17-6-4-3-5-7-17)12-21-26-13-15-8-10-16(11-9-15)20(24)25/h3-12H,13H2,1-2H3,(H,24,25)/b21-12-
Formula Weight 365.1376
Chemical Name or Material (Z)-Fenpyroximate (free acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.