missing translation for 'onlineSavingsMsg'
Learn More

Z-321, MedChemExpress

Product Code. 16490158
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490158 5 mg 5mg
16477698 10 mg 10mg
16401779 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490158 Supplier Medchem Express Supplier No. HY19123
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Z-321 is a prolylendopeptidase (PEP) inhibitor.

Z-321 is a prolylendopeptidase (PEP) inhibitor.

Specifications

Chemical Name or Material Z-321
CAS 130849-58-0
Color White
Purity Grade Notes Research
Molecular Formula C19H24N2O2S
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (725.75 mM; Need ultrasonic)
SMILES O=C([C@H]1N(C(CC2CC3=C(C=CC=C3)C2)=O)CSC1)N4CCCC4
Molecular Weight (g/mol) 344.47
Formula Weight 344.47
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.