missing translation for 'onlineSavingsMsg'
Learn More

(Z)-3-Hydroxy-9-octadecenoic Acid Ethyl Ester, TRC

Product Code. 30389671
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389671 100 mg 100mg
30404246 10 mg 10mg
30399101 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389671 Supplier Toronto Research Chemicals Supplier No. TRCH946715100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Z)-3-Hydroxy-9-octadecenoic Acid Ethyl Ester
Analyte or Component Names (Z)-3-Hydroxy-9-octadecenoic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C20H38O3
InChI Formula InChI=1S/C20H38O3/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-19(21)18-20(22)23-4-2/h11-12,19,21H,3-10,13-18H2,1-2H3/b12-11-
Solution Type Neat
SMILES CCCCCCCC/C=C\CCCCCC(O)CC(OCC)=O
IUPAC Name ethyl (Z)-3-hydroxyoctadec-9-enoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 326.51
Quantity 100 mg
Formula Weight 326.28
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.