missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)2S,4R-Sacubitril, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
(Z)2S,4R-Sacubitril is the impurity of Sacubitril. Sacubitril is approved by the Food and Drug Administration for use in combination with valsartan for the treatment of patients with heart failure.
Specifications
Specifications
| Chemical Name or Material | (Z)2S,4R-Sacubitril |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H27NO5 |
| Quantity | 5 mg |
| SMILES | [H]N(C(/C=C\C(O)=O)=O)[C@H](CC(C=C1)=CC=C1C2=CC=CC=C2)C[C@@H](C)C(OCC)=O |
| Molecular Weight (g/mol) | 409.47 |
| Formula Weight | 409.47 |
| Percent Purity | 96.89% |
| Grade | Research |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction