missing translation for 'onlineSavingsMsg'
Learn More

(Z)2S,4R-Sacubitril, MedChemExpress

Product Code. 16493469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493469 1 mg 1mg
16432728 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16493469 Supplier Medchem Express Supplier No. HYZ0075
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (Z)2S,4R-Sacubitril is the impurity of Sacubitril. Sacubitril is approved by the Food and Drug Administration for use in combination with valsartan for the treatment of patients with heart failure.

(Z)2S,4R-Sacubitril is the impurity of Sacubitril. Sacubitril is approved by the Food and Drug Administration for use in combination with valsartan for the treatment of patients with heart failure.

Specifications

Chemical Name or Material (Z)2S,4R-Sacubitril
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H27NO5
Quantity 1 mg
SMILES [H]N(C(/C=C\C(O)=O)=O)[C@H](CC(C=C1)=CC=C1C2=CC=CC=C2)C[C@@H](C)C(OCC)=O
Molecular Weight (g/mol) 409.47
Formula Weight 409.47
Percent Purity 96.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.