missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)-2-Nonen-1-ol, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | (Z)-2-Nonen-1-ol |
| CAS | 41453-56-9 |
| Molecular Formula | C9H18O |
| Quantity | 10 g |
| Synonym | (Z)-2-Nonenyl alcohol, cis-2-Nonen-1-ol, (2Z)-2-Nonen-1-ol |
| InChI Formula | InChI=1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h7-8,10H,2-6,9H2,1H3/b8-7- |
| SMILES | CCCCCC\C=C/CO |
| IUPAC Name | (Z)-non-2-en-1-ol |
| Molecular Weight (g/mol) | 142.24 |
| Formula Weight | 142.1358 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction