missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)-(2,4-Difluorophenyl)-(4-piperidyl)methanone Oxime, Mikromol™
Description
(Z)-(2,4-Difluorophenyl)-(4-piperidyl)methanone Oxime
Specifications
Specifications
| Chemical Name or Material | (Z)-(2,4-Difluorophenyl)-(4-piperidyl)methanone Oxime |
| CAS | 691007-05-3 |
| Analyte or Component Names | (Z)-(2,4-Difluorophenyl)-(4-piperidyl)methanone Oxime |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H14 F2 N2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Methanone, (2,4-difluorophenyl)-4-piperidinyl-, oxime, (1Z)-, (Z)-(2,4-Difluorophenyl)-4-piperidinylmethanone oxime |
| InChI Formula | InChI=1S/C12H14F2N2O/c13-9-1-2-10(11(14)7-9)12(16-17)8-3-5-15-6-4-8/h1-2,7-8,15,17H,3-6H2/b16-12- |
| SMILES | O\N=C(\C1CCNCC1)/c2ccc(F)cc2F |
| IUPAC Name | (NZ)-N-[(2,4-difluorophenyl)-piperidin-4-ylmethylidene]hydroxylamine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?