missing translation for 'onlineSavingsMsg'
Learn More

(Z)-2-(((1-(2-Aminothiazol-4-yl)-2-((2-hydroxy-2-sulfoethyl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic Acid, TRC

Product Code. 30348168
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348168 10 mg 10mg
30344483 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30348168 Supplier Toronto Research Chemicals Supplier No. TRCA73344510MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Z)-2-(((1-(2-Aminothiazol-4-yl)-2-((2-hydroxy-2-sulfoethyl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic Acid
CAS 2519572-35-9
Analyte or Component Names (Z)-2-(((1-(2-Aminothiazol-4-yl)-2-((2-hydroxy-2-sulfoethyl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic Acid
Shipping Condition Room Temperature
Molecular Formula C11H16N4O8S2
InChI Formula InChI=1S/C11H16N4O8S2/c1-11(2,9(18)19)23-15-7(5-4-24-10(12)14-5)8(17)13-3-6(16)25(20,21)22/h4,6,16H,3H2,1-2H3,(H2,12,14)(H,13,17)(H,18,19)(H,20,21,22)/b15-7-
Solution Type Neat
SMILES NC1=NC(/C(C(NCC(O)S(=O)(O)=O)=O)=N/OC(C)(C)C(O)=O)=CS1
IUPAC Name (Z)-2-(((1-(2-aminothiazol-4-yl)-2-((2-hydroxy-2-sulfoethyl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 396.4
Quantity 10 mg
Formula Weight 396.04
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.