missing translation for 'onlineSavingsMsg'
Learn More

(Z)-11-Octadecenoic Acid (Solution in Ethanol), TRC

Product Code. 30178936
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178936 50 mg 50mg
30179408 100 mg 100mg
30178929 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30178936 Supplier Toronto Research Chemicals Supplier No. TRCO86007550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-17-2
Molecular Formula C18 H34 O2
Molecular Weight (g/mol) 282.46
Synonym 11-Octadecenoic acid, (11Z)- (9CI, ACI), (11Z)-11-Octadecenoic acid (ACI), 11-Octadecenoic acid, (Z)- (8CI), 11-Octadecenoic acid, cis- (6CI), (Z)-11-Octadecenoic acid, (Z)-Octadec-11-enoic acid, 11-cis-Octadecenoic acid, 11-cis-Vaccenic acid, Asclepic acid, C18:1, cis-11-Octadecenoic acid, cis-11-Vaccenic acid, cis-Vaccenic acid, cis-Vaccenic acid, cis-δ11-Octadecenoic acid, δ11-cis-Octadecenoic acid
IUPAC Name (Z)-octadec-11-enoic acid
SMILES CCCCCC\C=C/CCCCCCCCCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h7-8H,2-6,9-17H2,1H3,(H,19,20)/b8-7-
Formula Weight 282.2559
Chemical Name or Material cis-Vaccenic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.